Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methylpyridin-3-ol 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Hydroxy-2-methylpyridine

B24944-06 B24944-14 B24944-03 B24944-14 B24944-03 B24944-06
AAAB24944-06EA 89.79 USD InStock
AAAB24944-06 CAAAAB24944-06 AAAB24944-14 AAAB24944-03 CAAAAB24944-14 CAAAAB24944-03
2-Methylpyridin-3-ol 99%
2-Methylpyridin-3-ol


Formula: C₆H₇NO
MW: 109.13 g/mol
Melting Pt: 166…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00082538
CAS Number: 1121-25-1
EINECS: 214-327-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty