Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Propoxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B24958-06 B24958-14 B24958-14 B24958-06
AAAB24958-06EA 33.89 USD InStock
AAAB24958-06 CAAAAB24958-06 AAAB24958-14 CAAAAB24958-14
4-Propoxybenzoic acid 98%
4-Propoxybenzoic acid


Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 145…147 °C
Density: 1,278 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013989
CAS Number: 5438-19-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty