Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Amino-4,6-dichloropyrimidine 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4,6-Dichloropyrimidin-5-ylamine

B24985-03 B24985-06 B24985-14 B24985-03 B24985-06 B24985-14
AAAB24985-06EA 190.99 USD InStock
CAAAAB24985-14 CAAAAB24985-03 CAAAAB24985-06 AAAB24985-06 AAAB24985-03 AAAB24985-14
5-Amino-4,6-dichloropyrimidine 98+%
5-Amino-4,6-dichloropyrimidine


Formula: C₄H₃Cl₂N₃
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 143…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00006108
CAS Number: 5413-85-4
EINECS: 226-503-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty