Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluorosalicylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Fluoro-2-hydroxybenzoic acid

B24992-06 B24992-03 B24992-14 B24992-03 B24992-06 B24992-14
AAAB24992-06EA 185.99 USD InStock
AAAB24992-06 AAAB24992-03 AAAB24992-14 CAAAAB24992-06 CAAAAB24992-14 CAAAAB24992-03
4-Fluorosalicylic acid 98%
4-Fluorosalicylic acid


Formula: C₇H₅FO₃
MW: 156.11 g/mol
Melting Pt: 186…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00153168
CAS Number: 345-29-9
EINECS: 206-459-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty