Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorocyclopentane 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cyclopentyl chloride

B25023-36 B25023-22 B25023-36 B25023-22
AAAB25023-22EA 58.69 USD InStock
CAAAAB25023-22 AAAB25023-22 CAAAAB25023-36 AAAB25023-36
Chlorocyclopentane 99%
Chlorocyclopentane


Formula: C₅H₉Cl
MW: 104.58 g/mol
Boiling Pt: 113…114 °C
Melting Pt: –94 °C
Density: 1.003
Flash Pt: 15 °C(59 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001360
CAS Number: 930-28-9
EINECS: 213-212-5
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty