Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Hexynoic acid 96%

Supplier: Thermo Scientific Chemicals
Danger

B25132-03 B25132-06 B25132-14 B25132-14 B25132-06 B25132-03
AAAB25132-06EA 216.25 USD InStock
AAAB25132-06 CAAAAB25132-06 AAAB25132-03 AAAB25132-14 CAAAAB25132-14 CAAAAB25132-03
5-Hexynoic acid 96%
5-Hexynoic acid


Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 73…74 °C/0.2 mm
Melting Pt: 41 °C
Density: 0.990
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066346
CAS Number: 53293-00-8
EINECS: 000-000-0
UN: 2922
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty