Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-4-methylpyridine 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Bromo-4-picoline

B25253-14 B25253-06 B25253-03 B25253-06 B25253-03 B25253-14
AAAB25253-14EA 406.99 USD InStock
CAAAAB25253-03 CAAAAB25253-14 AAAB25253-14 CAAAAB25253-06 AAAB25253-03 AAAB25253-06
2-Bromo-4-methylpyridine 96%
2-Bromo-4-methylpyridine


Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 87 °C/10 mm
Melting Pt: 218…228 °C
Density: 1.545
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00082590
CAS Number: 4926-28-7
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty