Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Nitrophthalonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Nitrobenzene-1,2-dicarbonitrile3-Nitro-1,2-dicyanobenzene3-Nitro-1,2-benzenedicarbonitrile

B25269-06 B25269-06 B25269-14 B25269-14
AAAB25269-06EA 76.89 USD InStock
CAAAAB25269-06 AAAB25269-06 AAAB25269-14 CAAAAB25269-14
3-Nitrophthalonitrile 99%
3-Nitrophthalonitrile


Formula: C₈H₃N₃O₂
Melting Pt: 162…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00191558
CAS Number: 51762-67-5
ADR: 6.1,I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty