Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Phenoxybenzonitrile 96%

Supplier: Thermo Scientific Chemicals
Warning

B25364-09 B25364-04 B25364-04 B25364-09
AAAB25364-04EA 39.19 USD InStock
AAAB25364-04 CAAAAB25364-09 CAAAAB25364-04 AAAB25364-09
4-Phenoxybenzonitrile 96%
4-Phenoxybenzonitrile


Formula: C₁₃H₉NO
MW: 195.22 g/mol
Melting Pt: 46…48 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00017346
CAS Number: 3096-81-9
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty