Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ω,3,4-Trichloroacetophenone

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2,3',4'-Trichloroacetophenone2-Chloro-1-(3,4-dichlorophenyl)ethanone3,4-Dichlorophenacyl chlorideα,3,4-Trichloroacetophenone

B25397-14 B25397-06 B25397-06 B25397-14
AAAB25397-06EA 51.59 USD InStock
CAAAAB25397-14 CAAAAB25397-06 AAAB25397-06 AAAB25397-14
ω,3,4-Trichloroacetophenone
ω,3,4-Trichloroacetophenone


Formula: C₈H₅Cl₃O
MW: 223.49 g/mol
Boiling Pt: 171…173 °C/15 mm
Melting Pt: 43…45 °C
Storage Temperature: Ambient
MDL Number: MFCD03970382
CAS Number: 42981-08-8
UN: 1759
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty