Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(4-methoxyphenyl) disulfide 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1,2-Bis(4-methoxyphenyl)disulfane1,2-Bis(4-methoxyphenyl)disulphane

B25399-06 B25399-06 B25399-03 B25399-03
AAAB25399-03EA 61.57 USD InStock
AAAB25399-03 CAAAAB25399-06 AAAB25399-06 CAAAAB25399-03
Bis(4-methoxyphenyl) disulfide 97%
Bis(4-methoxyphenyl) disulphide


Formula: C₁₄H₁₄O₂S₂
MW: 278.4 g/mol
Boiling Pt: 200 °C/3 mm
Melting Pt: 37…41 °C
Storage Temperature: Ambient
MDL Number: MFCD00041358
CAS Number: 5335-87-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty