Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methylisophthalic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B25496-03 B25496-06 B25496-03 B25496-06
AAAB25496-06EA 142.99 USD InStock
CAAAAB25496-06 AAAB25496-06 CAAAAB25496-03 AAAB25496-03
5-Methylisophthalic acid 98%
5-Methylisophthalic acid


Formula: C₉H₈O₄
MW: 180.16 g/mol
Melting Pt: 296…300 °C
Storage Temperature: Ambient
MDL Number: MFCD00013986
CAS Number: 499-49-0
EINECS: 207-881-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty