Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phloroglucinol, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 1,3,5-Trihydroxybenzene

B25502-18 B25502-30 B25502-18 B25502-30
AAAB25502-18EA 61.48 USD InStock
CAAAAB25502-18 AAAB25502-18 CAAAAB25502-30 AAAB25502-30
Phloroglucinol, anhydrous 98%
Phloroglucinol
Reagent for pentoses, pentosans and aldehydes. Used as a bone decalcifier in microscopy specimens


Formula: C₆H₆O₃
MW: 126.11 g/mol
Melting Pt: 215…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00002286
CAS Number: 108-73-6
EINECS: 203-611-2
Merck Index: 12,07482
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty