Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitro-o-anisic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B25523-03 B25523-06 B25523-03 B25523-06
AAAB25523-06EA 65.59 USD InStock
AAAB25523-06 CAAAAB25523-03 CAAAAB25523-06 AAAB25523-03
4-Nitro-o-anisic acid 98%
4-Nitro-o-anisic acid


Formula: C₈H₇NO₅
MW: 197.15 g/mol
Melting Pt: 145…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00216573
CAS Number: 2597-56-0
EINECS: 219-998-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty