Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-2-chloro-4-fluorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Bromo-3-chloro-1-fluorobenzene2-Chloro-4-fluorobromobenzene4-Bromo-3-chlorofluorobenzene

B25538-14 B25538-06 B25538-14 B25538-06
AAAB25538-06EA 90.39 USD InStock
AAAB25538-06 CAAAAB25538-14 CAAAAB25538-06 AAAB25538-14
1-Bromo-2-chloro-4-fluorobenzene 98%
1-Bromo-2-chloro-4-fluorobenzene


Formula: C₆H₃BrClF
MW: 209.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00142587
CAS Number: 110407-59-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty