Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl (4-fluorobenzoyl)acetate 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Methyl 4-fluorobenzoylacetateMethyl 3-(4-Fluorophenyl)-3-oxopropionate3-(4-Fluorophenyl)-3-oxopropionic Acid Methyl Ester(4-Fluorobenzoyl)acetic Acid Methyl Ester

B25546-09 B25546-03 B25546-03 B25546-09
AAAB25546-09EA 250.99 USD InStock
AAAB25546-09 CAAAAB25546-09 CAAAAB25546-03 AAAB25546-03
Methyl (4-fluorobenzoyl)acetate 95%
Methyl (4-fluorobenzoyl)acetate


Formula: C₁₀H₉FO₃
MW: 196.18 g/mol
Boiling Pt: 91 °C/5 mm
Density: 1.228
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000355
CAS Number: 63131-29-3
EINECS: 263-889-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty