Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,3-Dimethyl-2-oxobutyric acid ∼60% in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Trimethylpyruvic Acid3,3-Dimethyl-2-oxobutanoic acid

B25582-30 B25582-18 B25582-30 B25582-18
AAAB25582-18EA 61.69 USD InStock
AAAB25582-18 CAAAAB25582-30 AAAB25582-30 CAAAAB25582-18
3,3-Dimethyl-2-oxobutyric acid ∼60% in aqueous solution
3,3-Dimethyl-2-oxobutyric acid


Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00154352
CAS Number: 815-17-8
EINECS: 212-418-2
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty