Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Acetylbenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B25616-03 B25616-06 B25616-03 B25616-06
AAAB25616-06EA 334.99 USD InStock
AAAB25616-06 AAAB25616-03 CAAAAB25616-06 CAAAAB25616-03
3-Acetylbenzoic acid 98%
3-Acetylbenzoic acid


Formula: C₉H₈O₃
MW: 164.16 g/mol
Boiling Pt: 343 °C (1013 hPa)
Melting Pt: 162…166 °C
Density: 1,23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00045847
CAS Number: 586-42-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty