Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Benzyloxy-6-azaindole-3-carboxaldehyde 96%

Supplier: Thermo Scientific Chemicals
Warning

H50173-MD H50173-03 H50173-MD H50173-03
AAAH50173-03EA 370.99 USD InStock
CAAAAH50173-MD AAAH50173-03 CAAAAH50173-03 AAAH50173-MD
5-Benzyloxy-6-azaindole-3-carboxaldehyde 96%
5-Benzyloxy-6-azaindole-3-carboxaldehyde


Formula: C₁₅H₁₂N₂O₂
MW: 252.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191784
CAS Number: 56795-92-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty