Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-N,N-dimethylacetamide 97%

Supplier: Thermo Scientific Chemicals
Danger

H50229-MD H50229-MD H50229-03 H50229-03
AAAH50229-03EA 265.99 USD InStock
AAAH50229-03 CAAAAH50229-MD AAAH50229-MD CAAAAH50229-03
2-Chloro-N,N-dimethylacetamide 97%
2-Chloro-N,N-dimethylacetamide


Formula: C₄H₈ClNO
Boiling Pt: 98…100 °C/11 mm
Density: 1.182
Storage Temperature: Ambient
MDL Number: MFCD00039360
CAS Number: 2675-89-0
EINECS: 220-224-4
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty