Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Cyanobenzoic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

H50237-06 H50237-03 H50237-03 H50237-06
AAAH50237-03EA 45.09 USD InStock
CAAAAH50237-06 AAAH50237-03 AAAH50237-06 CAAAAH50237-03
2-Cyanobenzoic acid 96%
2-Cyanobenzoic acid


Formula: C₈H₅NO₂
MW: 147.13 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00045796
CAS Number: 3839-22-3
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty