Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-1,3-propanediamine 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: N-Boc-1,3-propanediaminetert-Butyl (3-aminopropyl)carbamatetert-Butyl N-(3-Aminopropyl)carbamateN-Boc-1,3-diaminopropaneN-(3-Aminopropyl)carbamic Acid tert-Butyl EsterN-1-Boc-1,3-diaminopropane·HClN-(tert-Butoxycarbonyl)-1,3-propanediamine

H50304-03 H50304-03 H50304-06 H50304-06
AAAH50304-06EA 308.99 USD InStock
AAAH50304-06 CAAAAH50304-06 AAAH50304-03 CAAAAH50304-03
N-Boc-1,3-propanediamine 95%
N-Boc-1,3-propanediamine


Formula: C₈H₁₈N₂O₂
MW: 174.244 g/mol
Melting Pt: ∼ 22 °C
Storage Temperature: Ambient
MDL Number: MFCD00210021
CAS Number: 75178-96-0
UN: 3259
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty