Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Phenyl-1H-pyrazole-4-carboxylic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

H50305-MD H50305-03 H50305-03 H50305-MD
AAAH50305-MDEA 88.29 USD InStock
AAAH50305-MD CAAAAH50305-MD AAAH50305-03 CAAAAH50305-03
1-Phenyl-1H-pyrazole-4-carboxylic acid 99%
1-Phenyl-1H-pyrazole-4-carboxylic acid


Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00514518
CAS Number: 1134-50-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty