Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Benzhydryl-N-methylamine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N-Methylbenzhydrylamine

H50327-03 H50327-06 H50327-06 H50327-03
AAAH50327-06EA 205.99 USD InStock
AAAH50327-06 CAAAAH50327-03 AAAH50327-03 CAAAAH50327-06
N-Benzhydryl-N-methylamine 98%
N-Benzhydryl-N-methylamine


Formula: C₁₄H₁₅N
MW: 197.28 g/mol
Melting Pt: 38…42 °C
Storage Temperature: Ambient
MDL Number: MFCD00467853
CAS Number: 14683-47-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty