Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-3-(trifluoromethyl)quinoxaline 96%

Supplier: Thermo Scientific Chemicals
Warning

H50549-06 H50549-03 H50549-06 H50549-03
AAAH50549-06EA 487.99 USD InStock
CAAAAH50549-03 AAAH50549-06 CAAAAH50549-06 AAAH50549-03
2-Chloro-3-(trifluoromethyl)quinoxaline 96%
2-Chloro-3-(trifluoromethyl)quinoxaline


Formula: C₉H₄ClF₃N₂
MW: 232.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00243891
CAS Number: 254732-51-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty