Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(4-Bromophenyl)-2-pyrrolidinone 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 1-(4-Bromophenyl)pyrrolidin-2-one

H50563-03 H50563-06 H50563-06 H50563-03
AAAH50563-03EA 114.99 USD InStock
CAAAAH50563-03 AAAH50563-03 CAAAAH50563-06 AAAH50563-06
1-(4-Bromophenyl)-2-pyrrolidinone 98%
1-(4-Bromophenyl)-2-pyrrolidinone


Formula: C₁₀H₁₀BrNO
MW: 240.10 g/mol
Boiling Pt: 98…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00138502
CAS Number: 7661-32-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty