Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Benzyl-1H-tetrazole 99%

Supplier: Thermo Scientific Chemicals

H50675-03 H50675-06 H50675-03 H50675-06
AAAH50675-06EA 305.25 USD InStock
AAAH50675-06 CAAAAH50675-03 AAAH50675-03 CAAAAH50675-06
5-Benzyl-1H-tetrazole 99%
5-Benzyl-1H-tetrazole


Formula: C₈H₈N₄
Storage Temperature: Ambient
MDL Number: MFCD00068730
CAS Number: 18489-25-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty