Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(2-Oxocyclohexyl)propanenitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

H50722-03 H50722-06 H50722-06 H50722-03
AAAH50722-06EA 173.99 USD InStock
CAAAAH50722-06 AAAH50722-06 CAAAAH50722-03 AAAH50722-03
3-(2-Oxocyclohexyl)propanenitrile 99%
3-(2-Oxocyclohexyl)propanenitrile


Formula: C₉H₁₃NO
Boiling Pt: 138…142 °C/10 mm
Density: 1.021
Storage Temperature: Ambient
MDL Number: MFCD00013797
CAS Number: 4594-78-9
EINECS: 224-991-6
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty