Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(3-Chloro-2-pyridinyl)piperazine 98%

Supplier: Thermo Scientific Chemicals
Warning

H50859-MD H50859-03 H50859-MD H50859-03
AAAH50859-MDEA 83.49 USD InStock
AAAH50859-MD CAAAAH50859-MD AAAH50859-03 CAAAAH50859-03
1-(3-Chloro-2-pyridinyl)piperazine 98%
1-(3-Chloro-2-pyridinyl)piperazine


Formula: C₉H₁₂ClN₃
MW: 197.67
Storage Temperature: Ambient
MDL Number: MFCD08061034
CAS Number: 87394-55-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty