Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(2-Fluorophenyl)-2-propylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

H50999-MD H50999-03 H50999-MD H50999-03
AAAH50999-MDEA 96.79 USD InStock
AAAH50999-MD AAAH50999-03 CAAAAH50999-MD CAAAAH50999-03
1-(2-Fluorophenyl)-2-propylamine 97%
1-(2-Fluorophenyl)-2-propylamine


Formula: C₉H₁₂FN
Storage Temperature: Ambient
MDL Number: MFCD01708100
CAS Number: 1716-60-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty