Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Azidotrimethylsilane 94%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Trimethylsilyl azide

L00173-22 L00173-06 L00173-22 L00173-14 L00173-14 L00173-06
AAAL00173-06EA 54.62 USD InStock
AAAL00173-06 AAAL00173-22 CAAAAL00173-22 AAAL00173-14 CAAAAL00173-06 CAAAAL00173-14
Azidotrimethylsilane 94%
Azidotrimethylsilane


Formula: C₃H₉N₃Si
Boiling Pt: 95…99 °C
Melting Pt: –95 °C
Density: 0.872
Flash Pt: 6 °C(42 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00001986
CAS Number: 4648-54-8
EINECS: 225-078-5
UN: 1992
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty