Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloroacrylic acid 96%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Chloroacrylicacid

L00350-04 L00350-04 L00350-09 L00350-09
AAAL00350-04EA 90.39 USD InStock
AAAL00350-04 CAAAAL00350-04 AAAL00350-09 CAAAAL00350-09
2-Chloroacrylic acid 96%
2-Chloroacrylic acid


Formula: C₃H₃ClO₂
MW: 106.51 g/mol
Melting Pt: 63…67 °C
Storage Temperature: Freezer
MDL Number: MFCD00014336
CAS Number: 598-79-8
EINECS: 209-953-9
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty