Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-tert-Butoxycarbonylimidazole 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N-Boc-imidazoletert-Butyl 1H-imidazole-1-carboxylate

L00506-14 L00506-06 L00506-14 L00506-06
AAAL00506-14EA 79.74 USD InStock
CAAAAL00506-06 AAAL00506-14 AAAL00506-06 CAAAAL00506-14
N-tert-Butoxycarbonylimidazole 98%
N-tert-Butoxycarbonylimidazole


Formula: C₈H₁₂N₂O₂
MW: 168.20 g/mol
Melting Pt: 44…48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014497
CAS Number: 49761-82-2
EINECS: 256-475-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty