Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl bromopyruvate ≥75%, tech.

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Bromopyruvic acid ethyl ester3-Bromo-2-oxopropionic acid ethyl esterEthyl 3-bromo-2-oxopropanoate

L00582-06 L00582-06 L00582-22 L00582-14 L00582-14 L00582-22
AAAL00582-22EA 283.99 USD InStock
CAAAAL00582-22 CAAAAL00582-06 AAAL00582-22 AAAL00582-06 CAAAAL00582-14 AAAL00582-14
Ethyl bromopyruvate ≥75%, tech.
Ethyl bromopyruvate
Ethyl 3-bromopyruvate is employed in a synthesis of thioxothiazolidines from primary amines and carbon disulfide


Formula: BrH₂CC(O)COOCH₂CH₃
MW: 195.01 g/mol
Boiling Pt: 98…100 °C/10 mm
Density: 1.555
Flash Pt: 98 °C(208 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000204
CAS Number: 70-23-5
EINECS: 200-729-6
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty