Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylglyoxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Benzoylformic acid2-Oxo-2-phenylacetic acid

L00648-14 L00648-06 L00648-14 L00648-06
AAAL00648-14EA 126.99 USD InStock
CAAAAL00648-06 AAAL00648-14 AAAL00648-06 CAAAAL00648-14
Phenylglyoxylic acid 98%
Phenylglyoxylic acid


Formula: C₈H₆O₃
MW: 150.13 g/mol
Boiling Pt: 147…151 °C/12 mm
Melting Pt: 62…67 °C
Density: 1,38
Storage Temperature: Ambient
MDL Number: MFCD00002575
CAS Number: 611-73-4
EINECS: 210-278-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty