Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-6-methylbenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

L00706-06 L00706-06 L00706-03 L00706-03
AAAL00706-03EA 35.09 USD InStock
CAAAAL00706-06 AAAL00706-03 CAAAAL00706-03 AAAL00706-06
2-Chloro-6-methylbenzonitrile 98%
2-Chloro-6-methylbenzonitrile


Formula: C₈H₆ClN
MW: 151.60 g/mol
Boiling Pt: 143 °C/38 mm
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00001782
CAS Number: 6575-09-3
EINECS: 229-499-5
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty