Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triphenylvinylphosphonium bromide 97%

Supplier: Thermo Scientific Chemicals
Warning

L00708-22 L00708-14 L00708-06 L00708-22 L00708-06 L00708-14
AAAL00708-06EA 67.39 USD InStock
AAAL00708-06 AAAL00708-14 AAAL00708-22 CAAAAL00708-22 CAAAAL00708-14 CAAAAL00708-06
Triphenylvinylphosphonium bromide 97%
Triphenylvinylphosphonium bromide


Formula: C₂₀H₁₈BrP
Melting Pt: 182…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00011807
CAS Number: 5044-52-0
EINECS: 225-740-3
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty