Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromopyruvic acid 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Bromo-2-oxopropionic acid3-Bromopyruvic acid

L00720-18 L00720-04 L00720-09 L00720-18 L00720-04 L00720-09
AAAL00720-18EA 355.99 USD InStock
CAAAAL00720-18 AAAL00720-18 CAAAAL00720-09 AAAL00720-09 CAAAAL00720-04 AAAL00720-04
Bromopyruvic acid 97%
Bromopyruvic acid


Formula: BrH₂CC(O)COOH
MW: 166.96 g/mol
Melting Pt: 75…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002587
CAS Number: 1113-59-3
EINECS: 214-206-5
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty