Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrabromomethane, (max. 6% H₂O) 98% (dry weight)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Carbon tetrabromide

L00821-22 L00821-14 L00821-36 L00821-36 L00821-14 L00821-22
AAAL00821-14EA 38.49 USD InStock
CAAAAL00821-22 AAAL00821-14 AAAL00821-36 AAAL00821-22 CAAAAL00821-36 CAAAAL00821-14
Tetrabromomethane, (max. 6% H₂O) 98% (dry weight)
Tetrabromomethane


Formula: Br₄C
MW: 331.63 g/mol
Boiling Pt: 190 °C
Melting Pt: 90…95 °C
Density: 3.420
Storage Temperature: Ambient
MDL Number: MFCD00000117
CAS Number: 558-13-4
EINECS: 209-189-6
UN: 2516
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty