Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Dinitrotoluene 97%

Supplier: Thermo Scientific Chemicals
Danger

L00856-06 L00856-06 L00856-14 L00856-14
AAAL00856-14EA 82.86 USD InStock
AAAL00856-14 CAAAAL00856-06 CAAAAL00856-14 AAAL00856-06
2,6-Dinitrotoluene 97%
2,6-Dinitrotoluene


Formula: C₇H₆N₂O₄
MW: 182.14 g/mol
Boiling Pt: 285 °C
Melting Pt: 58…65 °C
Flash Pt: 207 °C(404 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007158
CAS Number: 606-20-2
EINECS: 210-106-0
UN: 3454
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty