Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl isothiocyanatoformate 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Ethoxycarbonyl isothiocyanate

L00911-14 L00911-14 L00911-06 L00911-06
AAAL00911-06EA 74.99 USD InStock
AAAL00911-06 CAAAAL00911-14 AAAL00911-14 CAAAAL00911-06
Ethyl isothiocyanatoformate 97%
Ethyl isothiocyanatoformate


Formula: C₄H₅NO₂S
MW: 131.16 g/mol
Boiling Pt: 44…46 °C/10 mm
Density: 1.110
Flash Pt: 50 °C(122 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004814
CAS Number: 16182-04-0
EINECS: 240-318-9
UN: 2929
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty