Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromobenzhydrazide 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Bromobenzoic hydrazide2-Bromobenzene-1-carbohydrazide2-Bromobenzohydrazide2-Bromobenzoic acid hydrazide

L00937-14 L00937-06 L00937-06 L00937-14
AAAL00937-06EA 77.69 USD InStock
AAAL00937-06 CAAAAL00937-06 AAAL00937-14 CAAAAL00937-14
2-Bromobenzhydrazide 98+%
2-Bromobenzhydrazide


Formula: C₇H₇BrN₂O
MW: 215.05 g/mol
Melting Pt: 152…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00051577
CAS Number: 29418-67-5
EINECS: 249-614-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty