Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(1-Naphthoxy)acetic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(Naphthalen-1-yloxy)acetic acid

L01036-06 L01036-14 L01036-14 L01036-06
AAAL01036-06EA 45.49 USD InStock
AAAL01036-06 AAAL01036-14 CAAAAL01036-06 CAAAAL01036-14
(1-Naphthoxy)acetic acid 98+%
(1-Naphthoxy)acetic acid


Formula: C₁₂H₁₀O₃
MW: 202.21 g/mol
Melting Pt: 193…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00003927
CAS Number: 2976-75-2
EINECS: 221-023-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty