Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Iodo-p-xylene 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1,4-Dimethyl-2-iodobenzene2-Iodo-1,4-dimethylbenzene

L01055-09 L01055-18 L01055-09 L01055-18
AAAL01055-18EA 408.99 USD InStock
CAAAAL01055-18 CAAAAL01055-09 AAAL01055-18 AAAL01055-09
2-Iodo-p-xylene 98+%
2-Iodo-p-xylene


Formula: C₈H₉I
MW: 232.06 g/mol
Boiling Pt: 229…230 °C
Density: 1,617
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013708
CAS Number: 1122-42-5
EINECS: 214-347-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty