Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4,6-Tribromotoluene 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1,3,5-Tribromo-2-methylbenzene1-Methyl-2,4,6-tribromobenzene

L01149-18 L01149-09 L01149-09 L01149-18
AAAL01149-18EA 236.99 USD InStock
CAAAAL01149-09 CAAAAL01149-18 AAAL01149-18 AAAL01149-09
2,4,6-Tribromotoluene 98+%
2,4,6-Tribromotoluene


Formula: C₇H₅Br₃
MW: 328.83 g/mol
Boiling Pt: 290 °C
Melting Pt: 67…70 °C
Density: 2,479
Storage Temperature: Ambient
MDL Number: MFCD00013527
CAS Number: 6320-40-7
EINECS: 228-672-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty