Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromoacrylic acid 95%

Supplier: Thermo Scientific Chemicals
Danger

L01194-06 L01194-14 L01194-06
AAAL01194-06EA 69.69 USD InStock
AAAL01194-06 CAAAAL01194-06 AAL01194-14
2-Bromoacrylic acid 95%
2-Bromoacrylic acid


Formula: C₃H₃BrO₂
Melting Pt: 65…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014333
CAS Number: 10443-65-9
EINECS: 233-928-1
UN: 3261
ADR: "8",III
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty