Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9-Octadecynoic acid 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 9-ODA9-ODYA

L01294-06 L01294-06 L01294-03 L01294-03
AAAL01294-03EA 59.99 USD InStock
AAAL01294-03 CAAAAL01294-03 AAAL01294-06 CAAAAL01294-06
9-Octadecynoic acid 98%
9-Octadecynoic acid


Formula: C₁₈H₃₂O₂
MW: 280.45 g/mol
Boiling Pt: 189…190 °C/2 mm
Melting Pt: 43…47 °C
Storage Temperature: Ambient
MDL Number: MFCD00014386
CAS Number: 506-24-1
EINECS: 208-030-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty