Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cinnamyl bromide predominantly trans 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Bromo-1-phenyl-1-propene(3-Bromo-1-propen-1-yl)benzene

L01420-06 L01420-22 L01420-14 L01420-06 L01420-14 L01420-22
AAAL01420-06EA 36.89 USD InStock
AAAL01420-06 CAAAAL01420-22 AAAL01420-14 CAAAAL01420-14 AAAL01420-22 CAAAAL01420-06
Cinnamyl bromide predominantly trans 95%
Cinnamyl bromide


Formula: C₉H₉Br
MW: 197.08 g/mol
Boiling Pt: 88…90 °C/1 mm
Melting Pt: 26…29 °C
Flash Pt: 87 °C(188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000245
CAS Number: 4392-24-9
EINECS: 224-511-5
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty