Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluoro-DL-tyrosine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid

L01479-ME L01479-ME L01479-MC L01479-MC
AAAL01479-MEEA 317.99 USD InStock
AAAL01479-ME CAAAAL01479-ME AAAL01479-MC CAAAAL01479-MC
3-Fluoro-DL-tyrosine 97%
3-Fluoro-DL-tyrosine


Formula: C₉H₁₀FNO₃
MW: 199.18 g/mol
Melting Pt: ca 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00063075
CAS Number: 403-90-7
EINECS: 206-964-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty