Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Nitrothiophene-2-carboxaldehyde 98%

Supplier: Thermo Scientific Chemicals

L01493-14 L01493-06 L01493-06 L01493-14
AAAL01493-06EA 97.49 USD InStock
AAAL01493-06 CAAAAL01493-14 CAAAAL01493-06 AAAL01493-14
5-Nitrothiophene-2-carboxaldehyde 98%
5-Nitrothiophene-2-carboxaldehyde


Formula: C₅H₃NO₃S
MW: 157.15 g/mol
Melting Pt: 72…76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005433
CAS Number: 4521-33-9
EINECS: 224-850-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty